Gene Transcripts

Gene Symbol
PSORS1C1
Entrez Gene ID
170679
Gene Name
psoriasis susceptibility 1 candidate 1
Gene Aliases
C6orf16, SEEK1
Chromosome Location
Chr. 6: 31114831 - 31140092 on Build GRCh38
UniGene ID
Hs.310958
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014068.2 NP_054787.2 6 498
GenBank mRNA AB031479.1 - 6 483
AF484418.1 - 6 499
AF484419.1 - 2 104
BC104870.1 - 4 240 8143873
BI825917.1 - 3 160
DB454051.1 - 4 285

View Full Product Details