Gene Transcripts

Gene Symbol
OR10J5
Entrez Gene ID
127385
Gene Name
olfactory receptor family 10 subfamily J member 5
Gene Aliases
OR1-28
Chromosome Location
Chr. 1: 159535078 - 159536007 on Build GRCh38
UniGene ID
Hs.553587
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004469.1 NP_001004469.1 1 893
GenBank mRNA BC137025.1 - 1 938
BC137026.1 - 1 938 9021021

View Full Product Details