Gene Transcripts

Gene Symbol
OR2AT4
Entrez Gene ID
341152
Gene Name
olfactory receptor family 2 subfamily AT member 4
Gene Aliases
OR11-265
Chromosome Location
Chr. 11: 75088692 - 75100854 on Build GRCh38
UniGene ID
Hs.554515
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005285.1 NP_001005285.1 1 532
XM_017017655.1 XP_016873144.1 2 2172
XM_017017656.1 XP_016873145.1 2 2168
XM_017017657.1 XP_016873146.1 2 1710
XM_017017658.1 XP_016873147.1 2 2195
GenBank mRNA BC136958.1 - 1 573
BC136959.1 - 1 573 9020951

View Full Product Details