Gene Transcripts

Gene Symbol
UPF2
Entrez Gene ID
26019
Gene Name
UPF2 regulator of nonsense transcripts homolog (yeast)
Gene Aliases
HUPF2, RENT2, smg-3
Chromosome Location
Chr. 10: 11920022 - 12043170 on Build GRCh38
UniGene ID
Hs.370689
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015542.3 NP_056357.1 3 574
NM_080599.2 NP_542166.1 3 744
XM_011519449.2 XP_011517751.1 3 622
XM_017016034.1 XP_016871523.1 2 120
GenBank mRNA AB037829.1 - 2 943
AF301013.1 - 3 567
AF318574.1 - 3 544
AK096191.1 - 4 484
AK310925.1 - 3 533
AY013249.1 - 2 493
BC114964.1 - 3 538 40082739
BC115737.1 - 3 538 40082728

View Full Product Details