Gene Transcripts

Gene Symbol
TTC6
Entrez Gene ID
319089
Gene Name
tetratricopeptide repeat domain 6
Gene Aliases
C14orf25, NCRNA00291
Chromosome Location
Chr. 14: 37595199 - 37842628 on Build GRCh38
UniGene ID
Hs.659936
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001310135.1 NP_001297064.1 26 5460
XM_011537430.2 XP_011535732.1 22 3292
XM_011537431.2 XP_011535733.1 20 2949
XM_017021254.1 XP_016876743.1 25 5310
XM_017021255.1 XP_016876744.1 24 5175
XM_017021256.1 XP_016876745.1 25 5574
GenBank mRNA BC103914.2 - 7 809
BC103915.1 - 7 809 40006182
BC103916.2 - 7 809 40006183
BX161415.1 - 6 744

View Full Product Details