Gene Transcripts

Gene Symbol
Tbl3
Entrez Gene ID
287120
Gene Name
transducin (beta)-like 3
Gene Aliases
Chromosome Location
Chr. 10: 14066984 - 14072197 on Build Rnor_6.0
UniGene ID
Rn.32013
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008277.1 NP_001008278.1 13, 14 1373
GenBank mRNA BC085837.1 - 13, 14 1373

View Full Product Details