Gene Transcripts

Gene Symbol
LPAR4
Entrez Gene ID
2846
Gene Name
lysophosphatidic acid receptor 4
Gene Aliases
GPR23, LPA4, P2RY9, P2Y5-LIKE, P2Y9
Chromosome Location
Chr. X: 78747658 - 78758714 on Build GRCh38
UniGene ID
Hs.522701
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005296.2 NP_005287.1 2 2234
XM_005262126.3 XP_005262183.1 5 2652
XM_006724639.3 XP_006724702.1 4 2364
XM_017029437.1 XP_016884926.1 5 3711
XM_017029438.1 XP_016884927.1 3 2387
GenBank mRNA U90322.1 - 1 1895

View Full Product Details