Gene Transcripts

Gene Symbol
Krt40
Entrez Gene ID
287679
Gene Name
keratin 40, type I
Gene Aliases
Ka36
Chromosome Location
Chr. 10: 87453688 - 87459652 on Build Rnor_6.0
UniGene ID
Rn.140604
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008821.1 NP_001008821.1 7 1282

View Full Product Details