Gene Transcripts

Gene Symbol
Rqcd1
Entrez Gene ID
301513
Gene Name
Rcd1 required for cell differentiation1 homolog (S. pombe)
Gene Aliases
Chromosome Location
Chr. 9: 81783349 - 81808815 on Build Rnor_6.0
UniGene ID
Rn.18033
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009357.1 NP_001009357.1 8 3319
GenBank mRNA BC087134.1 - 8 3319

View Full Product Details