Gene Transcripts

Gene Symbol
PLEKHG4
Entrez Gene ID
25894
Gene Name
pleckstrin homology and RhoGEF domain containing G4
Gene Aliases
ARHGEF44, PRTPHN1, SCA4
Chromosome Location
Chr. 16: 67277820 - 67289500 on Build GRCh38
UniGene ID
Hs.188781
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001129727.2 NP_001123199.1 23 4338
NM_001129728.1 NP_001123200.1 22 4298
NM_001129729.2 NP_001123201.1 22 4271
NM_001129731.2 NP_001123203.1 20 4277
XM_011522985.2 XP_011521287.1 22 4268
XM_011522986.2 XP_011521288.1 22 4400
XM_011522987.2 XP_011521289.1 23 4344
XM_011522988.2 XP_011521290.1 22 4217
GenBank mRNA AB197663.1 - 21 6534
AB197664.1 - 20 6701
AK024475.1 - 22 4233
AL117435.1 - 17 3170
BC001520.2 - 7 1183
BC054486.1 - 20 4212 4820741
BC063501.1 - 22 4209 5527705
BC082974.1 - 23 4306 6291175

View Full Product Details