Gene Transcripts

Gene Symbol
Thap4
Entrez Gene ID
363291
Gene Name
THAP domain containing 4
Gene Aliases
Chromosome Location
Chr. 9: 100848597 - 100888107 on Build Rnor_6.0
UniGene ID
Rn.13958
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005564.1 NP_001005564.1 2 1124
GenBank mRNA BC081882.1 - 2 1124

View Full Product Details