Gene Transcripts

Gene Symbol
TBC1D2B
Entrez Gene ID
23102
Gene Name
TBC1 domain family member 2B
Gene Aliases
Chromosome Location
Chr. 15: 77994985 - 78077708 on Build GRCh38
UniGene ID
Hs.567426
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015079.5 NP_055894.6 9 1820
NM_144572.1 NP_653173.1 9 1820
XM_011521387.2 XP_011519689.1 8 1765
XM_017022022.1 XP_016877511.1 11 1841
GenBank mRNA AB028978.1 - 9 1683
AK000173.1 - 2 787
BC033712.1 - 4 603
BC140766.1 - 8 1465 9021634
BC144482.1 - 8 1465 9053009
BC152465.1 - 9 1683 8862601

View Full Product Details