Gene Transcripts

Gene Symbol
UBOX5
Entrez Gene ID
22888
Gene Name
U-box domain containing 5
Gene Aliases
RNF37, UBCE7IP5, UIP5, hUIP5
Chromosome Location
Chr. 20: 3107573 - 3159910 on Build GRCh38
UniGene ID
Hs.654646
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001267584.1 NP_001254513.1 5 2453
NM_014948.3 NP_055763.1 5 2458
NM_199415.2 NP_955447.1 4 2296
GenBank mRNA AB020667.1 - 5 2426

View Full Product Details