Gene Transcripts

Gene Symbol
UTP11
Entrez Gene ID
51118
Gene Name
UTP11, small subunit processome component homolog (S. cerevisiae)
Gene Aliases
CGI-94, CGI94, UTP11L
Chromosome Location
Chr. 1: 38012712 - 38024825 on Build GRCh38
UniGene ID
Hs.472038
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016037.3 NP_057121.2 3, 4 327
XM_011541555.2 XP_011539857.1 2, 3 261
GenBank mRNA AF067802.1 - 3, 4 290
AF151852.1 - 3, 4 305
AK023587.1 - 1, 2 1196
AK291900.1 - 3, 4 257
AK296018.1 - 3, 4 260
AK308377.1 - 2, 3 262
BC005182.1 - 3, 4 272 3995984
BU940926.1 - 3, 4 330

View Full Product Details