Gene Transcripts

Gene Symbol
ERGIC3
Entrez Gene ID
51614
Gene Name
ERGIC and golgi 3
Gene Aliases
C20orf47, CGI-54, Erv46, NY-BR-84, PRO0989, SDBCAG84, dJ477O4.2
Chromosome Location
Chr. 20: 35542029 - 35557634 on Build GRCh38
UniGene ID
Hs.472558
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015966.2 NP_057050.1 11 964
NM_198398.1 NP_938408.1 12 979
GenBank mRNA AF077030.1 - 11 922
AF091085.1 - 9 575
AF116614.1 - 11 1071
AF151812.1 - 11 922
AK074550.1 - 12 927
AL161963.1 - 9 620
BC009765.1 - 11 918 4283844
BC014014.2 - 11 917 3954269
EF206690.1 - 11 922

View Full Product Details