Gene Transcripts

Gene Symbol
TNFSF8
Entrez Gene ID
944
Gene Name
tumor necrosis factor superfamily member 8
Gene Aliases
CD153, CD30L, CD30LG, TNLG3A
Chromosome Location
Chr. 9: 114893343 - 114930595 on Build GRCh38
UniGene ID
Hs.494901
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001244.3 NP_001235.1 2, 3 538
NM_001252290.1 NP_001239219.1 2, 3 538
GenBank mRNA AK313619.1 - 2, 3 538
AL583443.3 - 2, 3 442
BC093630.1 - 2, 3 320 7939475
BC111939.1 - 2, 3 320 8327407
CD690231.1 - 2, 3 248
L09753.1 - 2, 3 360

View Full Product Details