Gene Transcripts

Gene Symbol
IRF2BPL
Entrez Gene ID
64207
Gene Name
interferon regulatory factor 2 binding protein like
Gene Aliases
C14orf4, EAP1
Chromosome Location
Chr. 14: 77024543 - 77028699 on Build GRCh38
UniGene ID
Hs.179260
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024496.3 NP_078772.1 1 1296
GenBank mRNA AB058768.1 - 2 791
AW001869.1 - 1 101
BC108292.1 - 1 1293
BC152431.1 - 2 791

View Full Product Details