Gene Transcripts

Gene Symbol
ODAM
Entrez Gene ID
54959
Gene Name
odontogenic, ameloblast asssociated
Gene Aliases
APIN
Chromosome Location
Chr. 4: 70195728 - 70204576 on Build GRCh38
UniGene ID
Hs.143811
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017855.3 NP_060325.3 6 513
XM_005265698.3 XP_005265755.1 7 577
XM_006714245.3 XP_006714308.1 7 554
XM_017008331.1 XP_016863820.1 5 469
GenBank mRNA AK000520.1 - 3 164
AK225547.1 - 3 144
BC017796.1 - 2 120 4689080
BG194365.1 - 6 516
EF113908.1 - 6 515

View Full Product Details