Gene Transcripts

Gene Symbol
Trim55
Entrez Gene ID
365751
Gene Name
tripartite motif-containing 55
Gene Aliases
Rnf29
Chromosome Location
Chr. 2: 104416934 - 104459117 on Build Rnor_6.0
UniGene ID
Rn.76142
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012218.1 NP_001012218.1 10 2420
XM_006232182.2 XP_006232244.1 9 2170
GenBank mRNA BC087100.1 - 10 2420

View Full Product Details