Gene Transcripts

Gene Symbol
PTRHD1
Entrez Gene ID
391356
Gene Name
peptidyl-tRNA hydrolase domain containing 1
Gene Aliases
C2orf79
Chromosome Location
Chr. 2: 24790267 - 24793382 on Build GRCh38
UniGene ID
Hs.406607
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013663.1 NP_001013685.1 1 107
GenBank mRNA BC073803.1 - 1 107

View Full Product Details