Gene Transcripts

Gene Symbol
Nhej1
Entrez Gene ID
363251
Gene Name
nonhomologous end-joining factor 1
Gene Aliases
Chromosome Location
Chr. 9: 82230230 - 82327923 on Build Rnor_6.0
UniGene ID
Rn.35483
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014217.1 NP_001014239.1 8 1401
XM_006245240.1 XP_006245302.1 8 1479
XM_006245241.2 XP_006245303.1 8 2055
XM_006245242.2 XP_006245304.1 8 1824
GenBank mRNA BC079033.1 - 8 1401

View Full Product Details