Gene Transcripts

Gene Symbol
SARS2
Entrez Gene ID
54938
Gene Name
seryl-tRNA synthetase 2, mitochondrial
Gene Aliases
SARS, SARSM, SERS, SYS, SerRS, SerRSmt, mtSerRS
Chromosome Location
Chr. 19: 38915264 - 38930896 on Build GRCh38
UniGene ID
Hs.709416
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017827.3 NP_060297.1 4 609
GenBank mRNA AB029948.1 - 4 609
AK000457.1 - 4 476
AK296152.1 - 4 473
BC042912.1 - 4 463 5478759

View Full Product Details