Gene Transcripts

Gene Symbol
TPD52L3
Entrez Gene ID
89882
Gene Name
tumor protein D52-like 3
Gene Aliases
NYDSP25, hD55
Chromosome Location
Chr. 9: 6328349 - 6331900 on Build GRCh38
UniGene ID
Hs.351815
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001874.2 NP_001001874.1 1 399
NM_001001875.3 NP_001001875.1 1 399
NM_033516.5 NP_277051.3 1 399
XM_017015280.1 XP_016870769.1 1 387
GenBank mRNA AK057479.1 - 1 373
AY032877.1 - 1 377
BC017589.2 - 1 373 4828082
BC042113.1 - 1 358 4838253

View Full Product Details