Gene Transcripts

Gene Symbol
SMU1
Entrez Gene ID
55234
Gene Name
DNA replication regulator and spliceosomal factor
Gene Aliases
BWD, SMU-1, fSAP57
Chromosome Location
Chr. 9: 33041852 - 33076716 on Build GRCh38
UniGene ID
Hs.655351
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018225.2 NP_060695.2 12 1571
XM_005251503.4 XP_005251560.1 11 1407
GenBank mRNA AK001667.1 - 12 1501
AK022032.1 - 4 375
AK225559.1 - 12 1526
AK225587.1 - 12 1520
AK304767.1 - 10 1158
BC002876.2 - 12 1517 3940669
BC110654.1 - 12 1611 6168993
EF011613.1 - 12 1517

View Full Product Details