Gene Transcripts

Gene Symbol
ZSCAN29
Entrez Gene ID
146050
Gene Name
zinc finger and SCAN domain containing 29
Gene Aliases
ZNF690, Zfp690
Chromosome Location
Chr. 15: 43358172 - 43371032 on Build GRCh38
UniGene ID
Hs.418287
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006720401.3 XP_006720464.1 1 443
XM_011521266.2 XP_011519568.1 1 443
XM_017021949.1 XP_016877438.1 1 443

View Full Product Details