Gene Transcripts

Gene Symbol
SLAIN2
Entrez Gene ID
57606
Gene Name
SLAIN motif family member 2
Gene Aliases
KIAA1458
Chromosome Location
Chr. 4: 48341480 - 48426198 on Build GRCh38
UniGene ID
Hs.479677
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020846.1 NP_065897.1 2 610
XM_005248121.2 XP_005248178.1 2 726
GenBank mRNA AB040891.1 - 2 580
AK026476.1 - 2 157
AK291006.1 - 2 740
BC040993.1 - 2 889 5272534

View Full Product Details