Gene Transcripts

Gene Symbol
TBC1D28
Entrez Gene ID
254272
Gene Name
TBC1 domain family member 28
Gene Aliases
Chromosome Location
Chr. 17: 18634434 - 18644431 on Build GRCh38
UniGene ID
Hs.434123
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039397.2 NP_001034486.2 10 1325
GenBank mRNA AK097563.1 - 9 1613
AL137747.1 - 1 152

Gene Symbol
TBC1D26
Entrez Gene ID
353149
Gene Name
TBC1 domain family member 26
Gene Aliases
Chromosome Location
Chr. 17: 15732277 - 15744784 on Build GRCh38
UniGene ID
Hs.434123
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK292544.1 - 9 1158
AK315847.1 - 9 1158
BC028414.1 - 8 1728

View Full Product Details