Gene Transcripts

Gene Symbol
Pcdhga12
Entrez Gene ID
498850
Gene Name
protocadherin gamma subfamily A, 12
Gene Aliases
Chromosome Location
Chr. 18: 30913655 - 30971113 on Build Rnor_6.0
UniGene ID
Rn.8628
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001037337.1 NP_001032414.1 1 1338
GenBank mRNA AY574011.1 - 1 1338

View Full Product Details