Gene Transcripts

Gene Symbol
Cyp4a10
Entrez Gene ID
13117
Gene Name
cytochrome P450, family 4, subfamily a, polypeptide 10
Gene Aliases
AI647584, Cyp4a, D4Rp1, RP1
Chromosome Location
Chr. 4: 115518287 - 115533649 on Build GRCm38
UniGene ID
Mm.10742
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AB017785.1 - 12, 13 1522
AB018421.1 - 12, 13 1558
AK098101.1 - 12, 13 1562
BC010747.1 - 12, 13 1549 4242518
BC051049.1 - 12, 13 1550 4234581

View Full Product Details