Gene Transcripts

Gene Symbol
Trp53i11
Entrez Gene ID
277414
Gene Name
transformation related protein 53 inducible protein 11
Gene Aliases
Tp53i11
Chromosome Location
Chr. 2: 93187534 - 93201757 on Build GRCm38
UniGene ID
Mm.41033
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025246.1 NP_001020417.1 8 2283
GenBank mRNA AK142316.1 - 9 2460
BC059041.1 - 7 2520
BC098206.1 - 8 2283 6812412

View Full Product Details