Gene Transcripts

Gene Symbol
Sh2d1b2
Entrez Gene ID
545378
Gene Name
SH2 domain containing 1B2
Gene Aliases
EAT-2B, Eat2b, Ert, Sh2d1c
Chromosome Location
Chr. 1: 170232871 - 170251587 on Build GRCm38
UniGene ID
Mm.379942
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033499.1 NP_001028671.1 3, 4 356
GenBank mRNA DQ131181.1 - 3, 4 356
DQ140178.1 - 3, 4 356

View Full Product Details