Gene Transcripts

Gene Symbol
SPRR2F
Entrez Gene ID
6705
Gene Name
small proline rich protein 2F
Gene Aliases
Chromosome Location
Chr. 1: 153112121 - 153119460 on Build GRCh38
UniGene ID
Hs.490252
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014450.2 NP_001014450.1 2 441
XM_017002176.1 XP_016857665.1 2 507

View Full Product Details