Gene Transcripts
- Gene Symbol
- NDUFV2
- Entrez Gene ID
- 4729
- Gene Name
- NADH:ubiquinone oxidoreductase core subunit V2
- Gene Aliases
- CI-24k
- Chromosome Location
- Chr. 18: 9102623 - 9134345 on Build GRCh38
- UniGene ID
- Hs.464572
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_021074.4 |
NP_066552.2
|
3, 4 |
307 |
|
XM_017025782.1 |
XP_016881271.1
|
3, 4 |
497 |
|
GenBank mRNA |
AK291865.1 |
- |
3, 4 |
203 |
|
AK312129.1 |
- |
3, 4 |
225 |
|
BC001632.1 |
- |
3, 4 |
217 |
3536345 |
BC017487.1 |
- |
3, 4 |
215 |
3537815 |
BC071689.1 |
- |
3, 4 |
197 |
4372795 |
BG742948.1 |
- |
3, 4 |
307 |
|
CB121843.1 |
- |
3, 4 |
260 |
|
M22538.1 |
- |
3, 4 |
211 |
|
View Full Product Details