Gene Transcripts

Gene Symbol
TCTA
Entrez Gene ID
6988
Gene Name
T-cell leukemia translocation altered
Gene Aliases
Chromosome Location
Chr. 3: 49412206 - 49416476 on Build GRCh38
UniGene ID
Hs.517962
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022171.2 NP_071503.1 3 767
GenBank mRNA AK000824.1 - 3 584
BC005157.1 - 3 733 2959904
L41143.1 - 3 767

View Full Product Details