Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_023012.5 | NP_075388.2 | 8 | 988 | |
NR_036434.1 | 9 | 1069 | |||
NR_036435.1 | 9 | 1088 | |||
NR_036436.1 | 8 | 1010 | |||
XM_005253601.1 | XP_005253658.1 | 8 | 991 | ||
XM_005253602.2 | XP_005253659.1 | 8 | 962 | ||
XM_005253604.1 | XP_005253661.1 | 9 | 1073 | ||
XM_011538688.1 | XP_011536990.1 | 8 | 991 | ||
XM_011538689.1 | XP_011536991.1 | 11 | 1246 | ||
XM_017019830.1 | XP_016875319.1 | 8 | 988 | ||
XM_017019831.1 | XP_016875320.1 | 9 | 1068 | ||
XM_017019832.1 | XP_016875321.1 | 11 | 1224 | ||
XM_017019833.1 | XP_016875322.1 | 9 | 1015 | ||
GenBank mRNA | AB212664.1 | - | 8 | 974 | |
AF161432.1 | - | 4 | 466 | ||
AK001883.1 | - | 8 | 948 | ||
AK023985.1 | - | 5 | 578 | ||
AK056114.1 | - | 9 | 1011 | ||
AK056256.1 | - | 8 | 948 | ||
BC008684.1 | - | 8 | 898 | 3853928 | |
BC067773.1 | - | 8 | 958 | 5301415 | |
BX640711.1 | - | 8 | 934 | ||
BX647884.1 | - | 8 | 977 |