Gene Transcripts

Gene Symbol
NOX5
Entrez Gene ID
79400
Gene Name
NADPH oxidase, EF-hand calcium binding domain 5
Gene Aliases
Chromosome Location
Chr. 15: 68930500 - 69057161 on Build GRCh38
UniGene ID
Hs.657932
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024505.3 NP_078781.3 3, 4 358
NR_033671.2 4, 5 589
GenBank mRNA AF325190.1 - 4, 5 564
AF353089.1 - 3, 4 337
AK074058.1 - 1, 2 27
AK314689.1 - 3, 4 337

View Full Product Details