Gene Transcripts
- Gene Symbol
- PKN1
- Entrez Gene ID
- 5585
- Gene Name
- protein kinase N1
- Gene Aliases
- DBK, PAK-1, PAK1, PKN, PKN-ALPHA, PRK1, PRKCL1
- Chromosome Location
- Chr. 19: 14433354 - 14471867 on Build GRCh38
- UniGene ID
- Hs.466044
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_002741.3 |
NP_002732.3
|
5, 6 |
932 |
|
NM_213560.1 |
NP_998725.1
|
5, 6 |
799 |
|
GenBank mRNA |
AK123007.1 |
- |
5, 6 |
813 |
|
AK292130.1 |
- |
5, 6 |
796 |
|
AK313886.1 |
- |
5, 6 |
772 |
|
BC040061.1 |
- |
5, 6 |
929 |
5752583 |
BC094766.1 |
- |
5, 6 |
963 |
30342125 |
D26181.1 |
- |
5, 6 |
800 |
|
S75546.1 |
- |
5, 6 |
848 |
|
U33053.1 |
- |
5, 6 |
848 |
|
View Full Product Details