Gene Transcripts

Gene Symbol
NUBPL
Entrez Gene ID
80224
Gene Name
nucleotide binding protein like
Gene Aliases
C14orf127, IND1, huInd1
Chromosome Location
Chr. 14: 31561385 - 31861293 on Build GRCh38
UniGene ID
Hs.288981
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025152.2 NP_079428.2 2 229
NR_120408.1 2 229
XM_011537182.2 XP_011535484.1 2 138
XM_011537183.2 XP_011535485.1 2 215
XM_017021664.1 XP_016877153.1 2 215
XM_017021665.1 XP_016877154.1 2 215
XM_017021666.1 XP_016877155.1 2 215
XM_017021667.1 XP_016877156.1 2 190
GenBank mRNA AK022722.1 - 2 142
AK295326.1 - 2 190
AK316445.1 - 2 219
BC024919.1 - 2 155 4475196
BP274976.1 - 2 229
BX248028.1 - 2 216
KJ534907.1 - 2 174
KJ534908.1 - 2 174
KJ534909.1 - 2 174

View Full Product Details