Gene Transcripts

Gene Symbol
WDR26
Entrez Gene ID
80232
Gene Name
WD repeat domain 26
Gene Aliases
CDW2, GID7, MIP2
Chromosome Location
Chr. 1: 224385143 - 224434299 on Build GRCh38
UniGene ID
Hs.497873
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001115113.2 NP_001108585.2 7 1271
NM_025160.6 NP_079436.4 7 1319
XM_005273273.4 XP_005273330.1 7 1220
XM_006711817.3 XP_006711880.1 7 1217
XM_011544281.2 XP_011542583.1 7 1060
XM_011544282.2 XP_011542584.1 4 382
XM_017002396.1 XP_016857885.1 7 908
GenBank mRNA AB209887.1 - 7 1319
AK024669.1 - 3 333
AK225061.1 - 3 333
AK299230.1 - 4 504
AY221751.1 - 7 1109
AY304473.1 - 7 876
BC031471.2 - 2 262
BC034498.1 - 8 1205
BC052301.1 - 6 715 6420398
BC063817.1 - 3 381
EF011612.1 - 6 714

View Full Product Details