Gene Transcripts
- Gene Symbol
- MARCH8
- Entrez Gene ID
- 220972
- Gene Name
- membrane associated ring-CH-type finger 8
- Gene Aliases
- CMIR, MARCH-VIII, MIR, RNF178, c-MIR
- Chromosome Location
- Chr. 10: 45454585 - 45594907 on Build GRCh38
- UniGene ID
- Hs.499489
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001002266.2 |
NP_001002266.1
|
2, 3 |
357 |
|
NM_001282866.1 |
NP_001269795.1
|
2, 3 |
357 |
|
NM_145021.5 |
NP_659458.2
|
2, 3 |
845 |
|
XM_005271804.2 |
XP_005271861.1
|
2, 3 |
846 |
|
XM_006717704.2 |
XP_006717767.1
|
2, 3 |
261 |
|
XM_011539492.2 |
XP_011537794.1
|
3, 4 |
897 |
|
XM_011539493.2 |
XP_011537795.1
|
4, 5 |
1138 |
|
XM_011539494.2 |
XP_011537796.1
|
3, 4 |
1087 |
|
XM_011539495.1 |
XP_011537797.1
|
2, 3 |
241 |
|
XM_017015894.1 |
XP_016871383.1
|
3, 4 |
897 |
|
GenBank mRNA |
AK057289.1 |
- |
1, 2 |
16 |
|
AK307667.1 |
- |
2, 3 |
357 |
|
AK313340.1 |
- |
2, 3 |
314 |
|
BC025394.2 |
- |
2, 3 |
334 |
4830278 |
BC066988.1 |
- |
2, 3 |
846 |
5267214 |
BF195460.1 |
- |
2, 3 |
538 |
|
BG717635.1 |
- |
2, 3 |
339 |
|
BM906478.1 |
- |
|
527 |
|
View Full Product Details