Gene Transcripts

Gene Symbol
C12orf54
Entrez Gene ID
121273
Gene Name
chromosome 12 open reading frame 54
Gene Aliases
HSD-29, HSD-30
Chromosome Location
Chr. 12: 48413154 - 48496514 on Build GRCh38
UniGene ID
Hs.98202
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152319.3 NP_689532.1 5, 6 292
XM_005268636.3 XP_005268693.1 4, 5 174
XM_011537896.2 XP_011536198.1 7, 8 940
XM_017018796.1 XP_016874285.1 9, 10 1130
GenBank mRNA AY255793.1 - 4, 5 197
BC031670.2 - 4, 5 218 5165130
BQ429029.1 - 5, 6 295

View Full Product Details