Gene Transcripts

Gene Symbol
RAB39B
Entrez Gene ID
116442
Gene Name
RAB39B, member RAS oncogene family
Gene Aliases
MRX72, WSMN
Chromosome Location
Chr. X: 155258234 - 155264589 on Build GRCh38
UniGene ID
Hs.632832
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_171998.3 NP_741995.1 2 3033
GenBank mRNA AK056209.1 - 1 1991
AL834460.1 - 2 2897

View Full Product Details