Gene Transcripts

Gene Symbol
SSNA1
Entrez Gene ID
8636
Gene Name
SS nuclear autoantigen 1
Gene Aliases
N14, NA-14, NA14
Chromosome Location
Chr. 9: 137188602 - 137190370 on Build GRCh38
UniGene ID
Hs.530314
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003731.2 NP_003722.2 3 558
GenBank mRNA BC000864.1 - 3 513
Z96932.1 - 3 479

View Full Product Details