Gene Transcripts

Gene Symbol
UBL5
Entrez Gene ID
59286
Gene Name
ubiquitin like 5
Gene Aliases
HUB1
Chromosome Location
Chr. 19: 9827880 - 9830121 on Build GRCh38
UniGene ID
Hs.534477
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001048241.2 NP_001041706.1 3, 4 259
NM_024292.3 NP_077268.1 3, 4 327
GenBank mRNA AF313915.1 - 3, 4 208
AK128885.1 - 3, 4 327
AK311889.1 - 3, 4 247
AV682189.1 - 3, 4 259
BC007053.1 - 3, 4 281 3827384
BC110839.1 - 3, 4 175 6474495
BP213482.1 - 3, 4 247
BT007355.1 - 2, 3 143

View Full Product Details