Gene Transcripts

Gene Symbol
NUBP2
Entrez Gene ID
10101
Gene Name
nucleotide binding protein 2
Gene Aliases
CFD1, NBP 2, NUBP1
Chromosome Location
Chr. 16: 1782923 - 1789191 on Build GRCh38
UniGene ID
Hs.256549
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001284501.1 NP_001271430.1 3, 4 428
NM_012225.3 NP_036357.1 2, 3 238
XM_005255027.2 XP_005255084.1 2, 3 705
XM_017022831.1 XP_016878320.1 2, 3 372
GenBank mRNA AF118394.1 - 2, 3 203
AK001023.1 - 2, 3 214
AK315611.1 - 2, 3 179
BC002768.2 - 2, 3 195 3633393
BC008005.1 - 2, 3 220 3507768
BU189242.1 - 2, 3 238

View Full Product Details