Gene Transcripts

Gene Symbol
SMCO4
Entrez Gene ID
56935
Gene Name
single-pass membrane protein with coiled-coil domains 4
Gene Aliases
C11orf75, FN5
Chromosome Location
Chr. 11: 93478472 - 93586856 on Build GRCh38
UniGene ID
Hs.438064
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020179.2 NP_064564.1 3 530
XM_011542907.1 XP_011541209.1 3 555
XM_011542908.2 XP_011541210.1 4 658
XM_011542909.1 XP_011541211.1 3 540
XM_011542910.2 XP_011541212.1 3 1293
XM_011542911.2 XP_011541213.1 3 1219
XM_017018019.1 XP_016873508.1 3 1275
XM_017018020.1 XP_016873509.1 4 716
GenBank mRNA AF197137.1 - 3 504
BC031564.1 - 3 530 5189479

View Full Product Details