Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001033925.1 | NP_001029097.1 | 6 | 1038 | |
NM_001323964.1 | NP_001310893.1 | 7 | 1363 | ||
NM_001323965.1 | NP_001310894.1 | 7 | 1106 | ||
NM_001323967.1 | NP_001310896.1 | 7 | 1055 | ||
NM_001323968.1 | NP_001310897.1 | 7 | 1100 | ||
NM_001323969.1 | NP_001310898.1 | 7 | 1049 | ||
NM_001323970.1 | NP_001310899.1 | 7 | 1100 | ||
NM_003252.3 | NP_003243.1 | 6 | 987 | ||
NR_136661.1 | 6 | 987 | |||
NR_136662.1 | 7 | 1106 | |||
NR_136663.1 | 7 | 1049 | |||
XM_017016606.1 | XP_016872095.1 | 3 | 991 | ||
GenBank mRNA | AB209260.1 | - | 2 | 1797 | |
AK290695.1 | - | 6 | 940 | ||
AK290984.1 | - | 6 | 637 | ||
AK291239.1 | - | 6 | 620 | ||
AK295243.1 | - | 7 | 500 | ||
AL833106.1 | - | 6 | 923 | ||
AY633610.1 | - | 6 | 431 | ||
BC030025.1 | - | 7 | 1368 | 4821759 | |
D64015.1 | - | 6 | 463 | ||
M96954.1 | - | 6 | 468 |