Gene Transcripts

Gene Symbol
Smim3
Entrez Gene ID
286910
Gene Name
small integral membrane protein 3
Gene Aliases
Nid67
Chromosome Location
Chr. 18: 55744809 - 55772354 on Build Rnor_6.0
UniGene ID
Rn.8865
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173126.3 NP_775149.1 2 633
XM_006254762.2 XP_006254824.1 4 1316
XM_006254763.2 XP_006254825.1 4 1319
XM_006254765.2 XP_006254827.1 4 845
XM_006254767.1 XP_006254829.1 2 319
XM_006254769.2 XP_006254831.1 2 522
XM_008772096.1 XP_008770318.1 3 1447
XM_008772097.1 XP_008770319.1 2 1255
GenBank mRNA AF313411.1 - 2 432
BC070515.1 - 2 633 7100997
EV780394.1 - 2 414

View Full Product Details