Gene Transcripts

Gene Symbol
Krt40
Entrez Gene ID
406221
Gene Name
keratin 40
Gene Aliases
Ka36
Chromosome Location
Chr. 11: 99537485 - 99543203 on Build GRCm38
UniGene ID
Mm.422372
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039666.1 NP_001034755.1 4 782
GenBank mRNA BC107359.1 - 4 782
BC107360.2 - 4 780
BC119522.2 - 4 782 40055959

View Full Product Details