Gene Transcripts

Gene Symbol
Zar1l
Entrez Gene ID
545824
Gene Name
zygote arrest 1-like
Gene Aliases
XM_359419
Chromosome Location
Chr. 5: 150502038 - 150518194 on Build GRCm38
UniGene ID
Mm.380068
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001159693.1 NP_001153165.1 1, 2 576
GenBank mRNA FJ858201.1 - 1, 2 576

View Full Product Details